Chemical-Resistant Multipurpose Trays and Pans
Actual product may vary
MFG Tray
Chemical-Resistant Multipurpose Trays and Pans
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W x H)Type
FIS15-239-718 x 14 x 1 in.Multi-purpose Tray
FIS15-239-1519.5 x 19.5 x 1.25 in.Multi-purpose Tray and Shallow Pan
FIS15-239-1624 x 12 x 2.75 in.Multi-purpose Tray and Ventilated Stacking Pan
FIS15-239-413.25 x 10.62 x 1 in.Multi-purpose Tray
FIS15-239-520.38 x 15.12 x 1 in.Multi-purpose Tray
Catalog number FIS15-239-7
Price (SGD)
105.00
Each
Dimensions (L x W x H):
18 x 14 x 1 in.
Type:
Multi-purpose Tray
Price (SGD)
105.00
Each
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
  • Reinforced white thermoset polyester composite construction Resistant to most detergents, mild acid and alkaline solutions, corrosive salts and many reagents
  • Withstand temperatures from -51° to +121°C (-60° to +250°F) continuous exposure, or 149°C (300°F) intermittent
  • High load-bearing capacity
  • Excellent dimensional stability Easily steam cleaned
Specifications
Dimensions (L x W x H)18 x 14 x 1 in.
ColorWhite
Height (Metric)2.5 cm
Length (Metric)46 cm
MaterialFiberglass
TypeMulti-purpose Tray
Width (Metric)35.5 cm
For Use With (Application)Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
Length (English)18 in.
Height (English)1 in.
ShapeRectangular
Temperature (English)-60°F to +250°F
Temperature (Metric)-51°C to +121°C
Width (English)14 in.
Unit SizeEach