Specifications
Product TypeMedium-duty Cloth
Length (English)13 in.
Length (Metric)33 cm
Width (English)14.4 in.
Width (Metric)31.75 cm
MaterialNon-woven Polyproylene Cellulose
AbsorbancySuper
ColorWhite
DisposableSingle-use
Dimensions (L x W)13 x 12.5 in. (33 x 31.75 cm)
For Use With (Application)Ideal for all manufacturing and maintenance and repair
Unit SizeCase