Chex-All™ II Instant-Seal Pouches
Chex-All™ II Instant-Seal Pouches
Actual product may vary
Chex-All™ II Instant-Seal Pouches
Chex-All™ II Instant-Seal Pouches
Propper
Chex-All™ II Instant-Seal Pouches
Self-Sealing Chex-All™ II Instant-Seal Pouches used in steam autoclaves or ethylene oxide sterilizers provide a barrier to maintain sterility after the sterilization process. Pre-printed indicators inside and outside the pouch. Triple-sealed paper/plastic film ensures content security.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Width (Metric)
FIS15-234-1538.1 cm12.7 cm
FIS15-234-1120.3 cm7.6 cm
FIS15-234-1330.5 cm17.8 cm
FIS15-234-1440.6 cm20.3 cm
FIS15-234-1225.4 cm12.7 cm
FIS15-234-1645.7 cm30.5 cm
CTL02-401-538.1 cm12.7 cm
Catalog number FIS15-234-15
Price (SGD)
954.00
Case
Add to cart
Length (Metric):
38.1 cm
Width (Metric):
12.7 cm
Price (SGD)
954.00
Case
Add to cart
Eliminate need for separate indicator strips
  • Self-Sealing Sterilization Pouches: Chex-All™ II sterilization pouches to contain instruments during steam and ethylene oxide (EO, EtO) sterilization.
  • Chex-All™ pouches feature pre-printed process indicators inside and outside the pouch.
  • The steam or ethylene oxide exposure will not affect the 3/4 in.wide adhesive line, guaranteeing a strong flat seal.
  • Self-Sealing Sterilization Pouches: Chex-All™ II is validated for double pouch packaging.
  • After steam sterilization, the chemical indicator changes from yellow to black.
  • After ethylene oxide sterilization, the chemical indicator changes from red to green.
  • Made of 2.5 mm plastic and 73-74 gram paper.
  • Triple sealed paper/plastic film provides excellent sterility protection.
  • Validated for double pouch packaging.
Specifications
AutoclavableAutoclavable
Closure TypeSelf-adhesive
ColorBlue
DisposableSingle-use
For Use With (Application)Indicators on both inside and outside for steam and ethylene oxide (EO) gas
MaterialPaper, Plastic
SterilityNon-sterile
No. per Case500/Cs.
Dimensions (L x W)15 x 5 in. (38.1 x 12.7 cm)
Length (English)15 in.
Width (English)5 in.
Length (Metric)38.1 cm
Width (Metric)12.7 cm
Unit SizeCase