Check Order Status
Glazed Handle Unglazed Head Porcelain Pestle
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Actual product may vary
Glazed Handle Unglazed Head Porcelain Pestle
Use the Porcelaines Avignon™ Glazed Handle Unglazed Head Porcelain Pestle. Its spherical mortar with beak is ideal for your suitable laboratory needs,
Have Questions?
Contact Us
Catalog Number
Length (Metric)
FIS15-207
170mm
Catalog number
FIS15-207
Price (SGD)
163.00
EA
Add to cart
Length (Metric):
170mm
Price (SGD)
163.00
EA
Add to cart
Scid resistant
Incompatible with hydrofluoric acid and Alkali metal salts
To be completed with pestle
Specifications
Finish
Glazed handle, unglazed head
Product Type
Pestle
Material
Porcelain
Length (Metric)
170mm
Unit Size
EA
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Safety Data Sheets
Share catalog number, name or link