Temperature Data Logger
Temperature Data Logger
Actual product may vary
Temperature Data Logger
1 million reading storage capacity
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCertifications/Compliance
FIS15-059-002CERCE, NIST
FIS15-059-002CE
Catalog number FIS15-059-002CER
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Certifications/Compliance:
CE, NIST
1 million reading storage capacity
Using the data logging software, starting, stopping and downloading from the unit is simple and easy. Graphical, tabular and summary data is provided for analysis and data can be viewed in °C, °F, K or °R. The data can also be automatically exported to Excel™ for further calculations. Free firmware upgrades for the life of the product are available, so that data loggers already deployed in the field can grow with the technology. Units do not need to be returned to the factory for upgrades. The user can do this automatically from any PC.
  • Multiple start/stop function
  • Ultra-high speed download capability
  • One million reading storage capacity
  • Optional memory wrap
  • Battery life indicator
  • Optional password protection
  • Programmable high-low alarms
  • 10 year battery life (at 15-min. reading rate, ambient temp. 25°C
Temperature
  • Temperature Sensor: Precision RTD Element
  • Temperature Range: -40°C to +80°C (-40°F to +176°C)
  • Temperature Resolution: 0.01°C (±0.018°F)
  • Temperature Calibrated: Accuracy ±0.5°C (±0.9°F)
General
  • Memory: 1,000,000 readings; software configurable memory wrap; 330,000 readings in multiple start/stop mode
  • Reading rate: One reading every second to every 24 hours
  • Baud rate: 115,200
  • LED indicators: Red and Green
  • Push-button start and stop
  • ABS plastic case
  • Dimensions: 1.4 x 2.2 x 0.6 in. (3.6 x 5.6 x 1.6cm)
  • Operating environment: -40ã+80°C (-40°F to +176°C), 0%RHã95% RH noncondensing
  • Required interface package: IFC200 (Catalog no. 15-060-036)
Specifications
TypeTemperature Datalogger
Data Memory1,000,000 Readings
CalibrationDigital Calibration
Alarm TypesHigh/Low, Selectable
Battery Type3.6 V Lithium
Battery Life10 Years
IncludesNIST calibration certificate
Certifications/ComplianceCE, NIST
InterfaceIFC200 Interface
MaterialABS Plastic
ModelTEMP101A
Sensor TypesPrecision RTD Element
For Use With (Application)For cold chain, refrigerator and freezer monitoring, vaccine monitoring, warehouse mapping, HVAC
Temperature Resolution0.01°C
Temperature Range (Metric)-40°C to +80°C
Operating Temperature (Metric)-40°C to +80°C
Weight (English)0.8 oz.
Weight (Metric)24 g
Unit SizeEA