PYREX™ Disposable Round-Bottom Rimless Glass Tubes
Actual product may vary
PYREX™ Disposable Round-Bottom Rimless Glass Tubes
Manufactured from borosilicate glass to reduce pH changes and contaminants. Corning™ PYREX™ Disposable Round-Bottom Rimless Glass Tubes are designed for both tissue culture and general bacteriological work.
Have Questions?
Catalog NumberCapacity (Metric)Length (Metric)
FIS14-962-1C6 mL75 mm
Catalog number FIS14-962-1C
Price (SGD)
200.00
Case
Capacity (Metric):
6 mL
Length (Metric):
75 mm
Price (SGD)
200.00
Case
Manufactured from borosilicate glass to reduce pH changes and contaminants. Corning™ PYREX™ Disposable Round-Bottom Rimless Glass Tubes are designed for both tissue culture and general bacteriological work.
These one-use culture tubes are packaged for your convenience in easy-to-open, easy-to-use packs. The three small sizes come in a dispenser arrangement, and the larger sizes are bulk-packed. Made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda-lime glass.
Specifications
Capacity (Metric)6 mL
Length (Metric)75 mm
MaterialGlass
DisposableSingle-use
For Use WithFor tissue culture work and general bacteriological use
Label TypeNone
PackagingBulk
RimmedNo
SterilityNon-sterile
GraduatedNo
Diameter (Metric)12 mm
Product TypeCulture Tube
Unit SizeCase