Disposable Borosilicate Glass Tubes with Threaded End
Actual product may vary
Disposable Borosilicate Glass Tubes with Threaded End
Available with or without marking spot
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Length (Metric)Label TypeThread Size
FIS14-959-35A16 mL125 mmMarking Spot15-415
FIS14-957-76A8 mL100 mmNone13-415
FIS14-959-35AA12 mL100 mmMarking Spot15-415
FKS1232727912 mL100 mmMarking Spot15-415
Catalog number FIS14-959-35A
Price (SGD)
839.00
Case
Capacity (Metric):
16 mL
Length (Metric):
125 mm
Label Type:
Marking Spot
Thread Size:
15-415
Price (SGD)
839.00
Case
Available with or without marking spot
  • Round bottom
  • Without caps
Note: Caps suitable for use with the culture tubes include the following: 
Black Phenolic Screw Thread Closures/PTFE/14B (05-569-4, 05-569-5, 06-421-20)
Screw Caps for Disposable Glass Tubes/polypropylene/linerless (03-340-77C, 03-340-77D, 03-340-77E) 
Specifications
Certifications/ComplianceASTM E438, Type I, Class B
Capacity (Metric)16 mL
Length (Metric)125 mm
MaterialBorosilicate Glass
DisposableSingle-use
For Use WithFor tissue culture work and general bacteriological use
Label TypeMarking Spot
PackagingTray
Thread Size15-415
SterilityNon-sterile
Diameter (Metric)16 mm
Closure TypeNot Included
Product TypeDisposable Culture Tube
Unit SizeCase