Search Thermo Fisher Scientific
| Catalog Number | Capacity (Metric) | Closure Type | Sterility | Packaging |
|---|---|---|---|---|
| FIS14-959-1B | 14 mL | Snap Cap | Sterile | Bag of 25, 500/Cs. |
| CRG352045 | 19 mL | Screw Cap | Sterile | Individually wrapped, 500/Cs. |
| FIS14-959-40B | 19 mL | Screw Cap | Sterile | Individually wrapped, 500/Cs. |
| FKS10694331 | 19 mL | Screw Cap | Sterile | Individually wrapped, 500/Cs. |
| FIS14-959-42 | 16 mL | Screw Cap | Sterile | Individually wrapped, 500/Cs. |
| FIS14-959-38B | 16 mL | Screw Cap | Sterile | Bag of 125, 1000/Cs. |
| FIS14-959-5 | 5 mL | Not Included | Non-sterile | Bulk, 1000/Cs. |
| FIS14-959-1A | 5 mL | Snap Cap | Sterile | Bag of 25, 500/Cs. |
| CRG352054 | 5 mL | Snap Cap | Sterile | Bag of 125, 1000/Cs. |
| FIS14-959-2A | 5 mL | Snap Cap | Sterile | Bag of 125, 1000/Cs. |
| FIS14-959-38C | 8 mL | Screw Cap | Sterile | Bag of 125, 1000/Cs. |
| CRG352052 | 5 mL | Not Included | Sterile | 1000/Cs. |
| FIS14-959-6 | 5 mL | Not Included | Sterile | 1000/Cs. |
| FKS10579511 | 5 mL | Not Included | Sterile | 1000/Cs. |
| FIS14-959A | 5 mL | Snap Cap | Sterile | Individually wrapped, 500/Cs. |
| FIS14-959B | 14 mL | Snap Cap | Sterile | Individually wrapped, 500/Cs. |
The foundation for consistent research results