Glue-Free Standard Cotton Swab, Wood Handle
Actual product may vary
Puritan
Glue-Free Standard Cotton Swab, Wood Handle
6 in. Standard cotton tip,with wood handle uses no glue used during construction which eliminates chemical migration. Ideal swab for cleaning and removing residues as well as applying medicines.
Have Questions?
Catalog NumberLength (Metric)
FIS14-959-10215.2 cm
Catalog number FIS14-959-102
Price (SGD)
-
Length (Metric):
15.2 cm
  • No glue adhesive on tip used during construction;
  • Used for clenaing and removing residues in controlled and industrial settings;
  • Can be used for cleaning wounds and applying medicines;
  • Ideal as a disk drive head cleaner;
  • Made in USA
Specifications
Certifications/ComplianceCE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
Diameter (English) Handle0.1 in.
Length (English)6 in.
For Use With (Application)Special construction eliminates migration of binder, cleaning lenses and optical devices
Product TypeSwab
Diameter (Metric) Handle2.5 mm
Length (Metric)15.2 cm
Length (English) Handle5.81 in.
Length (Metric) Handle14.7 cm
Length (English) Tip0.625 in.
Length (Metric) Tip1.5 cm
Diameter (Metric) Tip0.47 cm
Diameter (English) Tip0.18 in.
SterilityNon-sterile
Handle MaterialWood
Tip StyleStandard
Tip MaterialCotton
Unit SizePack