Kimble™ Plain Disposable Borosilicate Glass Tubes
Actual product may vary
DWK Life Sciences
Kimble™ Plain Disposable Borosilicate Glass Tubes
Suitable for a variety of applications and cell washing procedures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Length (Metric)Diameter (Metric)
FIS14-957-90E1 mL50 mm6 mm
FIS14-957-96K36 mL150 mm20 mm
FIS0334116 mL75 mm12 mm
FIS03-341-415 mL100 mm16 mm
FIS03341519 mL125 mm16 mm
FIS03-341-310 mL100 mm13 mm
Catalog number FIS14-957-90E
Price (SGD)
224.00
Case
Capacity (Metric):
1 mL
Length (Metric):
50 mm
Diameter (Metric):
6 mm
Price (SGD)
224.00
Case
Suitable for a variety of applications and cell washing procedures.
  • These premium quality tubes feature sturdy, uniform bottoms and consistent lengths 
  • The 10 x 75 and 12 x 75mm sizes are suitable for cell washing procedures
  • Tubes are packed in convenient trays for ease of use on bench tops and in drawers
  • The 6 x 50mm size is often referred to as a “Durham tube”
  • Trays are shrink-wrapped in plastic film, with tubes remaining in constant alignment for protection against in-transit breakage and contamination
  • No marking spot
  • Designed from ASTM Specification E890
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Specifications
Certifications/ComplianceUSP Type I, ASTM E 438 Type I, Class B
Capacity (Metric)1 mL
Length (Metric)50 mm
MaterialBorosilicate Glass
DisposableSingle-use
For Use WithExcellent chemical resistance against sodium leaching
Label TypeNone
PackagingTray
Diameter (Metric)6 mm
Product TypePlain Tube
Unit SizeCase