PYREX™ Disposable Round Bottom Threaded Culture Tubes, Without Marking Spot or Caps
PYREX™ Disposable Round Bottom Threaded Culture Tubes, Without Marking Spot or Caps
Actual product may vary
PYREX™ Disposable Round Bottom Threaded Culture Tubes, Without Marking Spot or Caps
PYREX™ Disposable Round Bottom Threaded Culture Tubes, Without Marking Spot or Caps
PYREX™ Disposable Round Bottom Threaded Culture Tubes, Without Marking Spot or Caps
Made from borosilicate glass for use with samples sensitive to leaching from plastic tubes. Corning™ Pyrex™ Round Bottom Threaded Culture Tubes are designed for tissue culture and general bacteriological work.
Have Questions?
Catalog NumberCapacity (Metric)Length (Metric)
FIS14-957-86D11.5 mL100 mm
Catalog number FIS14-957-86D
Price (SGD)
2,091.00
Case
Add to cart
Capacity (Metric):
11.5 mL
Length (Metric):
100 mm
Price (SGD)
2,091.00
Case
Add to cart
A round bottom tube without marking spot. Tubes are made of borosilicate glass and are sold without caps.
For caps, see Cat. No. 99999.
Specifications
Design TypeScrew neck
Capacity (Metric)11.5 mL
Length (Metric)100 mm
MaterialBorosilicate Glass
DisposableSingle-use
For Use WithIdeal for use with samples sensitive to leaching from plastic tubes
Label TypeNone
PackagingBulk
Thread Size15-415
SterilityNon-sterile
AutoclavableAutoclavable
GraduatedNo
Diameter (Metric)16 mm
Closure TypeNot Included
Product TypeCulture Tube
Unit SizeCase