1x1 image pixel for data collection
Kimble™ KIMAX™ Reusable Test Tubes with Beaded Rim and Marking Spot
Kimble™ KIMAX™ Reusable Test Tubes with Beaded Rim and Marking Spot
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Reusable Test Tubes with Beaded Rim and Marking Spot
Marking spot is durable fused-on white enamel
Have Questions?
Catalog NumberDiameter (Metric)Length (Metric)Capacity (Metric)
FIS14-956D15 mm125 mm16 mL
Catalog number FIS14-956D
Price (SGD)
219.00
PK
Add to cart
Diameter (Metric):
15 mm
Length (Metric):
125 mm
Capacity (Metric):
16 mL
Price (SGD)
219.00
PK
Add to cart
Marking spot is durable fused-on white enamel
  • Kimax KG-33 glass
  • With sturdy lip
  • Uniform wall thickness for maximum heat transfer, chemical resistance
Specifications
Certifications/ComplianceASTM E982, Type I
Design TypePlain Beaded Rim
Capacity (Metric)16 mL
Length (Metric)125 mm
MaterialBorosilicate Glass
Label TypeMarking Spot
Diameter (Metric)15 mm
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link