Sterile Plastic Culture Tubes: Clear Polystyrene
Sterile Plastic Culture Tubes: Clear Polystyrene
Actual product may vary
Sterile Plastic Culture Tubes: Clear Polystyrene
Sterile Plastic Culture Tubes: Clear Polystyrene
Sterile Plastic Culture Tubes: Clear Polystyrene
Sterile Plastic Culture Tubes: Clear Polystyrene
Ideal for tissue culture procedures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Length (Metric)Closure TypePackaging
FIS14-956-6C14 mL100 mmNot IncludedBulk - 8 Pks. of 125
FIS14-956-3C5 mL75 mmSnap CapBlister Packs
FIS14-956-3A5 mL75 mmNot IncludedBulk - 1 Pk. of 125
FIS14-956-5A125 mmScrew CapIndividually Wrapped, 500 Pks. of 1
FIS14-956-5B16 mL125 mmScrew CapBlister Packs
FIS14-956-5C19 mL150 mmScrew CapIndividually Wrapped, 500 Pks. of 1
FIS14-956-6A14 mL100 mmSnap CapIndividually Wrapped, 500 Pks. of 1
Catalog number FIS14-956-6C
Price (SGD)
-
Capacity (Metric):
14 mL
Length (Metric):
100 mm
Closure Type:
Not Included
Packaging:
Bulk - 8 Pks. of 125
Ideal for tissue culture procedures
  • Gamma-irradiated and tested for sterility; ready to use
  • No chance of residual contamination
  • All screw-cap tubes have outside threads and unconstricted necks, making filling and pipetting easier
  • All snap caps can be positioned for aerobic or anaerobic applications
Specifications
ColorClear
Capacity (Metric)14 mL
Length (Metric)100 mm
MaterialPolystyrene
For Use WithIdeal for tissue culture procedures
PackagingBulk - 8 Pks. of 125
SterilitySterile
Diameter (Metric)17 mm
Closure TypeNot Included
Product TypeCulture Tube
Unit SizeCS