Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Length (Metric) | Closure Type | Packaging |
---|---|---|---|---|
FIS14-956-3A | 5 mL | 75 mm | Not Included | Bulk - 1 Pk. of 125 |
FIS14-956-3C | 5 mL | 75 mm | Snap Cap | Blister Packs |
FIS14-956-5A | 125 mm | Screw Cap | Individually Wrapped, 500 Pks. of 1 | |
FIS14-956-5B | 16 mL | 125 mm | Screw Cap | Blister Packs |
FIS14-956-5C | 19 mL | 150 mm | Screw Cap | Individually Wrapped, 500 Pks. of 1 |
FIS14-956-6A | 14 mL | 100 mm | Snap Cap | Individually Wrapped, 500 Pks. of 1 |
FIS14-956-6C | 14 mL | 100 mm | Not Included | Bulk - 8 Pks. of 125 |