Glass Dropper
Actual product may vary
Glass Dropper
Glass droppers used to transfer liquid
Have Questions?
Catalog NumberVolume (Metric)Tip Style
FIS14-955-5012.5 mLStraight
Catalog number FIS14-955-501
Price (SGD)
117.00
Pack
Volume (Metric):
2.5 mL
Tip Style:
Straight
Price (SGD)
117.00
Pack
Glass droppers used to transfer liquid
  • Choice of straight or curve design
  • Material: Glass
  • Choice of 2mL or 2.5mL volume
Specifications
MaterialGlass
Tip StyleStraight
Volume (English)0.084 oz.
Volume (Metric)2.5 mL
TypeDropper
Unit SizePack