Ultra Thin Magnetic Stirrer
Actual product may vary
Ultra Thin Magnetic Stirrer
Spill safe, maintenance free slim profile magnetic stirrer with pulse mode.
Have Questions?
Catalog NumberCapacity
FIS14-955-150800 mL
Catalog number FIS14-955-150
Price (SGD)
-
Capacity:
800 mL
Spill safe, maintenance free slim profile magnetic stirrer with pulse mode.
  • Modern magnetic coil technology
  • Flat surface
  • Compact and thin design
  • Pulse function provides better mixing (stir bar alternates rotating clockwise and counter clockwise every 30 seconds)
  • Simple control panel with easy-to-use interface
  • Variable speed setting from 15 to 1500rpm
  • Speed indicator during operation
  • Anti-skid pads
  • Wear-free drive (no moving parts)
  • Setup plate and casing made from chemically-resistant materials
  • Stir bar docking location to place or attach magnetic stir bar
Technical Specifications
  • Maximum stirring volume (H2O): 800mL
  • Maximum stirring capability (tested in H2O): 500mL at 1500rpm with 2.5cm stir bar
  • Speed range: 15 to 1500rpm
  • Max. stirring bar length: 2.5cm (0.98 in.)
  • Set-up plate material: Polyester
  • Set-up plate dimensions: 9.4cm (3.7 in.)
  • Dimensions (L x W x H): 18.5 x 11.5 x 1.3cm (7.2 x 4.5 x 0.5 in.)
  • Weight: 0.4 to 0.5Kg (0.8 to 1.2 lb.)
  • Permissible ambient temperature: 5 to 40°C
  • Protection class according to DIN EN 60529: IP 65
  • Input voltage: 100 to 230V
  • Frequency: 50/60Hz
  • Power: 5w
  • Auto direction change/Pulse Mode: Yes
  • Auto direction change time interval: 30 seconds
  • Current speed indicator: Yes
Specifications
Speed15 to 1500 rpm
Capacity800 mL
Voltage100/230 V
Wattage5 w
Dimensions (L x W x H)7.28 x 4.52 x 0.5 in. (18.5 x 11.5 x 1.3 cm)
Height (English)0.5 in.
Height (Metric)1.3 cm
Length (English)7.28 in.
Length (Metric)18.5 cm
Top Plate Length (English)3.7 in.
Top Plate Length (Metric)9.4 cm
Width (English)4.52 in.
Width (Metric)11.5 cm
Electrical Requirements100 to 230V 50/60Hz
Frequency50/60 Hz
ModelUltra Thin
Temperature Range (Metric)5°C to 40°C
Weight (English)0.8 to 1.2 lb.
Weight (Metric)0.4 to 0.5Kg
Unit SizeEach