Safe-T-Fill™ Capillary Blood Collection Systems: Lithium Heparin
Actual product may vary
RAM Scientific
Safe-T-Fill™ Capillary Blood Collection Systems: Lithium Heparin
No scooping needed, allowing for high-quality, tissue-free samples
Have Questions?
Catalog NumberCapacity (Metric)Additive
FIS14-915-69300 μLLithium Heparin with Gel
Catalog number FIS14-915-69
Price (SGD)
-
Capacity (Metric):
300 μL
Additive:
Lithium Heparin with Gel
No scooping needed, allowing for high-quality, tissue-free samples
  • Pierceable attached cap with self-sealing versions
  • Anticoagulant coated capillary and tube to prevent clotting
  • Expiration date and lot number on tube for quality control
Specifications
ColorClear
Product TypeChemistry/Plasma/Whole Blood Tube
AdditiveLithium Heparin with Gel
MaterialPlastic
SterilityNon-sterile
Closure TypeStandard Pierceable Cap
Capacity (Metric)300 μL
Closure IncludedYes
Closure ColorGreen
For Use With (Application)Intended for chemistry determinations that require plasma or whole blood samples. Gel separator serves as a barrier between blood and serum after centrifugation.
Unit SizePack