Search Fisher Scientific
| Catalog Number | Capacity (Metric) | Additive | Color |
|---|---|---|---|
| FIS14-915-59 | 200 μL | Clot Activator and Gel | Clear |
Blood samples are drawn into the SAFE-T-FILL™ Plastic Micro Capillary Blood Collection Tubes through capillary action, reducing hemolysis, tissue contamination, and re-draws. No scooping or milking required. Coated with Sodium Fluoride for glucose determinations.