Disposable Safelet I.V. Catheters
Actual product may vary
Exel International
Disposable Safelet I.V. Catheters
Precision taper assures smooth venipuncture with no catheter tip fold-back
Have Questions?
Catalog NumberNeedle GaugeColorDiameter (English) OuterLength (English)
FIS14-841-2124 GRed0.017 in.0.75 in.
Catalog number FIS14-841-21
Price (SGD)
369.00
Pack
Needle Gauge:
24 G
Color:
Red
Diameter (English) Outer:
0.017 in.
Length (English):
0.75 in.
Price (SGD)
369.00
Pack
Precision taper assures smooth venipuncture with no catheter tip fold-back
  • Functional casing is designed for one-handed opening with no rolling
  • Hydrophobic membrane eliminates blood leakage
  • Translucent hub allows easy detection of flashback
  • With PTFE Radio-opaque catheter
  • Color coded for easy gauge selection
  • Sterile, nontoxic, nonpyrogenic
  • Latex-free
Specifications
Certifications/CompliancePyrogen Free
MaterialPTFE
ColorRed
DescriptionHydrophobic Membrane filter
Needle Gauge24 G
Diameter (English) Outer0.017 in.
Diameter (Metric) Outer0.045 cm
For Use With (Application)Safe and smooth venipuncture. Eliminates blood residuals and helps reduce blood coagulation or hemolysis with its crimped smooth engagement.
Latex FreeYes
Length (English)0.75 in.
Length (Metric)2.11 cm
Product TypeWingless IV Catheter
DisposableSingle-use
Quantity50 Box
Connection TypeLuer Taper