Super-ject™ Dental Needles
Actual product may vary
Exel International
Super-ject™ Dental Needles
Plastic hub design allows for greater needle flexibility and lowers chances of needle breakage
Have Questions?
Catalog NumberNeedle GaugeLength (English) NeedleColor
FIS14-840-1230 G1.2 in.Blue
Catalog number FIS14-840-12
Price (SGD)
79.00
Pack
Needle Gauge:
30 G
Length (English) Needle:
1.2 in.
Color:
Blue
Price (SGD)
79.00
Pack
  • Red dot on the needle hub marks the needle bevel position, even when the needle point is not visible
  • Dental needle production is computer controlled in all processes from manufacture to inspection
  • Siliconized cannula
  • Needles are sterilized by Ethylene Oxide
  • Dental needle hubs and boxes are color-coded by gauge for easier identification
  • Self-tapping ribs placed on the outside of the hub, and inside threads, make for ease of use
  • Larger cannula lumen (.0055 in.)
Specifications
MaterialPlastic (Hub)
ColorBlue
Needle Gauge30 G
For Use With (Application)For greater needle flexibility and lowers chances of needle breakage
Length (English) Needle1.2 in.
Product TypeDental Needle
Length (Metric) Needle32 mm