Sharps Disposal Containers
Actual product may vary
Covidien
Sharps Disposal Containers
Choice of capacities and type of lid
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)Capacity (Metric)Closure TypeDimensions (L x W x H)
FIS14-830-1242 gal.7.6 LRotor Lid7.25 x 10.5 x 10 in.
FIS14-830-1328 gal.30.3 LHinged Top11 x 15.5 x 17.5 in.
Catalog number FIS14-830-124
Price (SGD)
329.00
Case
Capacity (English):
2 gal.
Capacity (Metric):
7.6 L
Closure Type:
Rotor Lid
Dimensions (L x W x H):
7.25 x 10.5 x 10 in.
Price (SGD)
329.00
Case
Choice of capacities and type of lid
Specifications
Capacity (Metric)7.6 L
Depth (English)7.25 in.
Closure TypeRotor Lid
Capacity (English)2 gal.
ColorRed
Dimensions (L x W x H)7.25 x 10.5 x 10 in.
Height (Metric)25.4 cm
Length (Metric)18.4 cm
Width (Metric)26.7 cm
Width (English)10.5 in.
Length (English)7.25 in.
Height (English)10 in.