▸
First Aid and Medical
▸
Sharps Disposal Containers and Accessories
▸
Multipurpose Sharps Containers
Actual product may vary
Covidien
Multipurpose Sharps Containers
Economical horizontal drop maximizes container volume
Have Questions?
Catalog Number
Capacity (English)
Closure Type
Color
FIS14-830-113
0.5 gal.
Lockable w/ Rotor Lid
Clear
Catalog number
FIS14-830-113
Price (SGD)
-
Capacity (English):
0.5 gal.
Closure Type:
Lockable w/ Rotor Lid
Color:
Clear
Large horizontal openings accommodate larger syringes
Containers lock for final disposal
Specifications
Capacity (Metric)
1.9 L
Depth (English)
4.75 in.
Closure Type
Lockable w/ Rotor Lid
Capacity (English)
0.5 gal.
Color
Clear
For Use With (Application)
Needle syringe collection
Width (English)
4.75 in.
Length (English)
4.75 in.
Height (English)
4.5 in.