Search Thermo Fisher Scientific
| Catalog Number | Capacity (English) | Capacity (Metric) | Closure Type | Shape |
|---|---|---|---|---|
| FIS14-827-105 | 7.5 gal. | 28 L | Lockable | Round |
| FIS14-827-104 | 7 gal. | 26.5 L | Lockable | Rectangle |
| FIS14-827-63 | 1 gal. | 3.8 L | Lockable | Rectangle |
| FIS22-037-959 | 2 gal. | 7.5 L | Slide Top | Rectangle |