▸
First Aid and Medical
▸
Patient Care Products
▸
Medical Syringes
▸
Laboratory Pipetting Needles
Actual product may vary
Laboratory Pipetting Needles
316 Stainless Steel Hub with 316 Stainless Steel Tubing
Have Questions?
Catalog Number
Needle Gauge
Length (English) Needle
Material
Connection Type
FIS14-825-16M
14 G
4 in.
Nickel-plated (Hub), Stainless Steel
Luer
Catalog number
FIS14-825-16M
Price (SGD)
844.00
Pack
Add to cart
Needle Gauge:
14 G
Length (English) Needle:
4 in.
Material:
Nickel-plated (Hub), Stainless Steel
Connection Type:
Luer
Price (SGD)
844.00
Pack
Add to cart
316 Stainless Steel Hub with 316 Stainless Steel Tubing
Needles 14-825-16A through -16N and 14-818-106A through -106C all have standard nickel plated hubs and 304 stainless steel cannulae
Needles 01-290-29 through -35 have 316 stainless steel Micro-Mate™ Threaded Female Luer Hubs and 316 stainless steel cannulae
All have 90° blunt ends
Specifications
Material
Nickel-plated (Hub), Stainless Steel
Needle Gauge
14 G
For Use With (Application)
For both manual and automatic volumetric dispensers, diluters, pipetters and for laboratory procedures
Length (English) Needle
4 in.
Product Type
Pipetting Needle
Quantity
12 Needles
Needle Point Style
90° Blunt
Connection Type
Luer