Disposable Syringes with Luer-Lok™ Tips
Actual product may vary
Fisherbrand
Disposable Syringes with Luer-Lok™ Tips
Unique integrated BD Luer-Lok™ tip syringe with clear barrel. BD Medical™ Disposable Syringes with Luer-Lok™ Tips are sterile and disposable.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
FIS14-823-301 mL
FIS14-829-455 mL
FIS14-823-4353 mL
Catalog number FIS14-823-30
Price (SGD)
-
Volume (Metric):
1 mL
Unique integrated BD Luer-Lok™ tip syringe with clear barrel. BD Medical™ Disposable Syringes with Luer-Lok™ Tips are sterile and disposable.
  • Sterile syringe for single use
  • Without needle
  • Clear barrel (polycarbonate or polypropylene) with bold scale markings
  • Tapered plunger rod for ease of aspiration
  • Positive plunger rod stop
  • Added BD Luer-Lok™ thread for increased secure connection
  • DNR (Latex) free
  • Shelf life: 1825 days
Specifications
Product TypeSyringe
DisposableSingle-use
Needle Point NumberNo Needle Included
Volume (Metric)1 mL
Connection TypeLuer Lock