Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
First Aid and Medical
▸
Patient Care Products
▸
Medical Syringes
▸
HSW Soft-Ject™ Disposable Syringes
Actual product may vary
Air-Tite
HSW Soft-Ject™ Disposable Syringes
High-quality, three-part disposable rubber piston syringes
Have Questions?
Change view
Catalog Number
Volume (Metric)
FIS14-817-57
50 mL
FIS14-817-54
10 mL
FIS14-817-53
5 mL
3 Options
Catalog number
FIS14-817-57
Price (SGD)
-
Volume (Metric):
50 mL
High-quality, three-part disposable rubber piston syringes
Lubricated synthetic rubber piston allows for smooth draw while positive stop helps prevent accidental pullout
Bold graduations are easily read and resist wipe off
ETO gas sterilized syringes, latex and pyrogen free
Packaged in individual sterile strip packs
Made in Germany
Specifications
Certifications/Compliance
FDA, CE, ISO 9001
Latex Free
Yes
Product Type
Soft-Ject™ Syringe
Disposable
Single-use
Graduations
60 mL
Quantity
Pk. of 50
Volume (Metric)
50 mL
Connection Type
Luer Lock
Sterility
Sterile