Search Fisher Scientific
| Catalog Number | Volume (Metric) | Graduations | Connection Type |
|---|---|---|---|
| FIS14-817-33 | 20 mL | 1 cc | Luer Tip |
| FIS14-817-27 | 3 mL | 0.1 cc | Luer Lock |
| FIS14-817-36 | 50 mL | 2 cc | Catheter |
| FIS14-817-40 | 50 mL | 2 cc | Luer Tip |
| FIS14-817-41 | 30 mL | 1 cc | Luer Tip |
| FIS14-817-25 | 1 mL | 0.05 cc | Leur Slip |
| FIS14-817-26 | 3 mL | 0.1 cc | Centric Luer |
| FIS14-817-31 | 10 mL | 0.5 cc | Luer Tip |
| FIS14-817-28 | 5 mL | 0.2 cc | Centric Luer Slip |
| FIS14-817-32 | 20 mL | 1 cc | Eccentric Luer Tip |
| FIS14-817-30 | 10 mL | 0.5 cc | Eccentric Luer Tip |
| FIS14-817-35 | 50 mL | 1 cc | Eccentric Luer Tip |
| FIS14-817-44 | 10 mL | 0.05 cc | Centric Luer Slip |