All-Plastic Henke-Ject™ Syringes
All-Plastic Henke-Ject™ Syringes
Actual product may vary
Air-Tite
All-Plastic Henke-Ject™ Syringes
Inert syringes for use in gas or liquid chromatography, in vitro fertilization (IVF), embryo-transfer, and various laboratory procedures. Air-Tite All Plastic Henke-Ject™ syringes are available compatible with a variety of needle styles. Needles are sold separately.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)GraduationsConnection Type
FIS14-817-3220 mL1 ccEccentric Luer Tip
FIS14-817-273 mL0.1 ccLuer Lock
FIS14-817-3320 mL1 ccLuer Tip
FIS14-817-3650 mL2 ccCatheter
FIS14-817-4050 mL2 ccLuer Tip
FIS14-817-4130 mL1 ccLuer Tip
FIS14-817-251 mL0.05 ccLeur Slip
FIS14-817-263 mL0.1 ccCentric Luer
FIS14-817-3110 mL0.5 ccLuer Tip
FIS14-817-285 mL0.2 ccCentric Luer Slip
FIS14-817-3010 mL0.5 ccEccentric Luer Tip
FIS14-817-3550 mL1 ccEccentric Luer Tip
FIS14-817-4410 mL0.05 ccCentric Luer Slip
Catalog number FIS14-817-32
Price (SGD)
142.00
Pack
Add to cart
Volume (Metric):
20 mL
Graduations:
1 cc
Connection Type:
Eccentric Luer Tip
Price (SGD)
142.00
Pack
Add to cart
These Air-Tite, all plastic syringes are available compatible with a variety of needle styles. (Needles are sold separately.) Ideal for use in gas or liquid chromatography, in vitro fertilization (IVF), embryo-transfer, and various laboratory procedures.
  • Needle tips not included and must be purchased separately
  • Graduated syringes manufactured from high-quality laboratory-grade polypropylene and polyethylene
  • Chemically inert and noncytotoxic, these syringes are ideal for chromatography, in vitro fertilization and embryo transfer
  • Luer Tip, Luer-Slip, or catheter tip compatibility
  • Polypropylene barrel and polyethylene plunger are chemically inert and noncytotoxic; needles contain no latex, synthetic rubber, silicone oil, or styrene
  • Positive safety stop prevents accidental spills
  • FDA registered
Specifications
Certifications/ComplianceFDA, CE, ISO 9001, DEHP Free
MaterialPlastic
For Use With (Application)For nuclear medicine, amniocentisis, IVF, embryo-transfer, chromatography and many laboratory procedures
Latex FreeYes
Product TypeHenke-Ject™ Syringe
Graduations1 cc
Volume (Metric)20 mL
Product LineNorm-Ject
Needle Point StyleCone
Connection TypeEccentric Luer Tip