1000 Series Gastight™ Syringes: Removable-Needle, RN Termination, 22 Gauge, 2 in. Needle Length
Actual product may vary
Hamilton
1000 Series Gastight™ Syringes: Removable-Needle, RN Termination, 22 Gauge, 2 in. Needle Length
Mid-volume GASTIGHT syringe ideal for dispensing volumes from 100 μL up to 100 mL.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Needle Point NumberNeedle Point Style
FIS14-815-1675 mL2Sharp, Bevel, Curved, Non-coring
FIS14-815-18310 mL2Sharp, Bevel, Curved, Non-coring
Catalog number FIS14-815-167
Price (SGD)
-
Volume (Metric):
5 mL
Needle Point Number:
2
Needle Point Style:
Sharp, Bevel, Curved, Non-coring
Mid-volume GASTIGHT syringe ideal for dispensing volumes from 100 μL up to 100 mL.
Removable Needle (RN): the needles are removable and are a Hamilton-specific design. The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length and point style to optimize the syringe for custom applications. Additionally, this termination allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life.
Needle Point Styles
  • Style 2: Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 33-10
  • Style 3: Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 33-10
Specifications
AutoclavableAutoclavable
MaterialBorosilicate Glass (Barrel), PTFE (Tip), Stainless Steel (Plunger)
Needle Gauge22 G
Diameter (English) Inner0.41 in.
Diameter (English) Inner Needle0.016 in.
Diameter (Metric) Inner Needle0.41 mm
Diameter (English) Outer Needle0.028 in.
Diameter (English) Outer0.53 in.
Diameter (Metric) Inner10.3 mm
Diameter (Metric) Outer13.5 mm
Diameter (Metric) Outer Needle0.72 mm
For Use With (Application)Ideal for manual and automated HPLC injections, commonly used in syringe pumps and other liquid handling instruments
Length (English) Needle2 in.
Product TypeGastight Syringe
Max. Pressure200 psi
Max. Temperature (English)239°F
Max. Temperature (Metric)115°C
Min. Temperature (English)50°F
Model1005
Needle Point Number2
QuantityEa.
Volume (Metric)5 mL
Needle Point StyleSharp, Bevel, Curved, Non-coring
Connection TypeRemovable
Unit SizeEach