AutoclavableAutoclavable
ColorBlue
HoldsTubes (13 to 16 mm)
MaterialEpoxy-coated Steel
Positions48
ShapeRectangular
StackableNo
SterilityNon-sterile
Dimensions (L x W x H)9.5 x 3.6 x 2.5 in. (24.2 x 9.2 x 6.4 cm)
Length (Metric)24.2 cm
Length (English)9.5 in.
Width (English)3.6 in.
Height (English)2.5 in.
Diameter (Metric) Opening16 mm
Product TypeTest Tube Rack
Diameter (Metric) Inner16 mm
Shelves3 Tiers
Height (Metric)6.4 cm
Width (Metric)9.2 cm
Unit SizeEach