Rodent Liquid Diet, Lieber-DeCarli 82, Shake and Pour
Actual product may vary
Bio-Serv
Rodent Liquid Diet, Lieber-DeCarli 82, Shake and Pour
No blender needed—just add water, shake, and pour.
Have Questions?
Catalog NumberProduct Type
FIS14-726-531Rodent Liquid Diet (Control)
Catalog number FIS14-726-531
Price (SGD)
-
Product Type:
Rodent Liquid Diet (Control)
No blender needed—just add water, shake, and pour
  • Easy to prepare
  • Offers the versatility of a liquid format, allowing the addition of alcohol, medications, or test compounds
  • Provides a convenient way to measure food intake or study nutritional manipulation
  • Ideal for post-operative recovery
  • Nutritionally assayed
  • Powder
  • Ingredients: Corn Syrup Solids, Casein, Olive Oil, Maltodextrin, Cellulose, Mineral Mix, Corn Oil, Suspending Aids, Safflower Oil, Vitamin Mix, L-Cystine, Choline Bitartrate, DL-Methionine
Specifications
Product TypeRodent Liquid Diet (Control)
Calories1000
ComponentsCorn syrup solids, Casein, Olive Oil, Maltodextrin, Cellulose, Mineral Mix, Corn Oil, Suspending aids, Safflower Oil, Vitamin Mix, L-Cystine, Choline Bitartrate, DL-Methionine
FormatLiquid
For Use With (Application)Ideal for post operative recovery
Particle ShapePowder
Quantity4 L
Shelf Life6 Months
SpeciesRodent
Storage RequirementsRefrigerate
Unit SizeEach