Mouse Igloo™ Rodent Enrichment Device, Certified
Actual product may vary
Bio-Serv
Mouse Igloo™ Rodent Enrichment Device, Certified
Provides shelter and a secluded place to build a nest
Have Questions?
Catalog NumberColor
FIS14-726-502Amber
Catalog number FIS14-726-502
Price (SGD)
-
Color:
Amber
Provides shelter and a secluded place to build a nest
  • Transparent material provides ease of visibility
  • Great addition to breeding programs
  • Fits most shoebox cages
  • Stacks easily for convenient storage
  • Made of high temperature polycarbonate
  • Autoclavable
  • Cagewasher safe
  • Certified (Contaminant screened)
Specifications
AutoclavableAutoclavable
Product TypeMouse Igloo
ColorAmber
MaterialPolycarbonate
For Use With (Application)Provide shelter and a secluded place to build a nest, great addition to breeding programs
Height (English)2.25 in.
Height (Metric)5.7 cm
Unit SizeEach