NLK Protein, Active
Product Image
Bel-Art
NLK Protein, Active
Active, N-terminal 6His-tagged, recombinant, human NLK, amino acids 121-end, for use in Kinase Assays.
Have Questions?
Catalog NumberQuantity
FIS14-67610 μg
Catalog number FIS14-676
Price (SGD)
445.00
EA
Add to cart
Quantity:
10 μg
Price (SGD)
445.00
EA
Add to cart
NLK (Nemo Like Kinase) regulates a number of transcription factors with key roles in cell fate determination. It enables ubiquitin protein ligase binding activity and Iinvolved in protein stabilization and transforming growth factor beta receptor signaling pathway. It predicted to be located in cytosol and nucleoplasm, and to be active in cytoplasm and nucleus.
GenBank NM_016231
Specifications
Formulationmg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol)48.7 kDa
Quantity10 μg
SpeciesHuman
RecombinantRecombinant
Protein TagN-terminal 6His
Expression SystemRecombinant enzyme expressed in Sf21 insect cells
Protein FormTruncated
Purity or Quality Grade≥53%
ProteinNLK, active
Unit SizeEA