Low-Cost Cast-Iron Tripod-Base Supports
Low-Cost Cast-Iron Tripod-Base Supports
Actual product may vary
Low-Cost Cast-Iron Tripod-Base Supports
Finished with chemical-resistant, baked, black enamel
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) RodDiameter (Metric) RodLength (English) RodLength (Metric) Rod
FIS14-675DQ0.5 in.1.3 cm36 in.91 cm
FIS14-675BQ0.37 in.0.95 cm20 in.51 cm
FIS14-675CQ0.43 in.1.1 cm24 in.61 cm
Catalog number FIS14-675DQ
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (English) Rod:
0.5 in.
Diameter (Metric) Rod:
1.3 cm
Length (English) Rod:
36 in.
Length (Metric) Rod:
91 cm
Finished with chemical-resistant, baked, black enamel
  • Base is heavy, rugged cast iron for stability
  • Support rods and base precisely threaded to eliminate instability
Specifications
MaterialCast Iron
Length (English) Rod36 in.
Length (Metric) Rod91 cm
Diameter (English) Rod0.5 in.
Diameter (Metric) Rod1.3 cm
Product TypeTripod Base Support
FinishBaked Black Enamel
For Use With (Application)Support rods and base precisely threaded to eliminate instability
Quantity1 Ea.
Unit SizeEA