Microcentrifuge Tubes with Locking Snap Cap
Microcentrifuge Tubes with Locking Snap Cap
Actual product may vary
Fisherbrand
Microcentrifuge Tubes with Locking Snap Cap
Polypropylene microcentrifuge tubes are DNase/RNase and pyrogen free
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSterilityCapacity (Metric)Max. RCF
FIS14-666-313NaturalSterile2.0 mL25,000 x g
FIS14-666-315NaturalNon-sterile2.0 mL25,000 x g
FKS15442545NaturalNon-sterile2.0 mL25,000 x g
FIS14-666-318NaturalSterile1.5 mL26,000 x g
FIS14-666-319NaturalNon-sterile1.5 mL26,000 x g
FKS15432545NaturalNon-sterile1.5 mL26,000 x g
FIS14-666-323RedNon-sterile1.5 mL26,000 x g
FIS14-666-332NaturalSterile0.5 mL30,000 x g
Catalog number FIS14-666-313
Price (SGD)
6.00
Pack
Add to cart
Color:
Natural
Sterility:
Sterile
Capacity (Metric):
2.0 mL
Max. RCF:
25,000 x g
Price (SGD)
6.00
Pack
Add to cart
Polypropylene microcentrifuge tubes are DNase/RNase and pyrogen free
  • DNase/RNase and pyrogen free
  • Available in both Sterile and Nonsterile
  • Autoclavable and withstand temperatures to -86°C
  • Graduated tubes available in three sizes - 0.5mL, 1.5mL and 2.0mL
Specifications
Cap StyleLocking snap cap
AutoclavableAutoclavable
Capacity (Metric)2.0 mL
ColorNatural
MaterialPolypropylene
PackagingIndividually wrapped
SterilitySterile
GraduatedYes
Max. RCF25,000 x g
Diameter (Metric)13 mm
Length (Metric)40 mm
Unit SizePack