5 mL Polypropylene Snaplock Microcentrifuge Tubes
Actual product may vary
Axygen
5 mL Polypropylene Snaplock Microcentrifuge Tubes
For use in low volume sample requirements
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSterility
FIS14-568-100ClearNon-sterile
FKS15227179ClearNon-sterile
Catalog number FIS14-568-100
Price (SGD)
298.00
Pack
Color:
Clear
Sterility:
Non-sterile
Price (SGD)
298.00
Pack
Axygen MaxyClear microcentrifuge tubes are available in a variety of colors. Axygen Maxymum Recovery™ surface and sterile options are also available. SnapLock microcentrifuge tubes have frosted cap surfaces and frosted side panels, ideal for labeling. Each cap features a piercing port in the center of the cap. Microcentrifuge tubes with no caps are available for some sizes.
  • 0.6 mL, 1.5 mL, and 2.0 mL clear microcentrifuge tubes are boil proof. Colored tubes are not boil-proof.
  • Co-polymer tubes offer exceptional strength and chemical resistance. Not to be autocalvable or boil-proof.
  • Clear microcentrifuge tubes are boil-proof. Colored tubes are not boil-proof.
  • Maximum G-force is 14,000 RCF.
  • RNase-/DNase-free and nonpyrogenic
  • Sterility Assurance Level (SAL) of 10−6
  • Color - Assorted: Each pack contains a mixture of blue, green, violet, orange, red, and yellow.
  • Clear microcentrifuge tubes are boil-proof. Colored tubes are not boil-proof.
  • Color - Spectrum: 2 packs each of blue, green, orange, red, and yellow.
Specifications
Cap StyleSnap Cap
Closure MaterialPolypropylene
Capacity (Metric)5 mL
ColorClear
MaterialHomopolymer
No. per Pack250/Pk.
SterilityNon-sterile
GraduatedYes
Closure TypeSnap Cap
Bottom ShapeCylindrical
Product TypeMicrocentrifuge Tube
Unit SizePack