Waring™ Two-Speed 1L Blenders
Actual product may vary
Waring™ Two-Speed 1L Blenders
Waring™ Two-Speed 1L Blenders
Conair
Waring™ Two-Speed 1L Blenders
Two-speed heavy-duty laboratory blenders
Have Questions?
Change viewbuttonViewtableView
Catalog NumberContainer Material
FIS14-509-53Glass
FIS14-509-54Stainless Steel
Catalog number FIS14-509-53
Price (SGD)
2,099.00
Each
Container Material:
Glass
Price (SGD)
2,099.00
Each
  • Die-Cast Zinc Plated Base - with baked enamel coating for durability and good looks
  • Open Blender Base Collar- lets spills drain away from the motor base
  • Heavy Duty Blending Assembly with 4 Integral Blades - Stainless stesl blades hold up under continual use, ensuring consistent blending
  • Speed: 18,000 and 22,000rpm
  • 120VAC, 50/60Hz
  • 7D x 8W x 16.5 in. H
Specifications
Amperage3 A
Speed18,000, 22,000 rpm
Voltage120 V
Electrical Requirements120 V 50/60 Hz
Hertz50/60 Hz
Horsepower0.5 hp
Wattage350 W
Capacity (Metric)1000 mL
Sample Volume (English)33.81 oz.
Depth (English)7 in.
Depth (Metric)17.78 cm
Height (English)16.5 in.
Height (Metric)41.9 cm
Width (English)8 in.
Width (Metric)20.32 cm
Base MaterialDie Cast Zinc
Container MaterialGlass
IncludesHandle and two piece vinyl and styrene lid
Packaging Quantity1 Ea.
Unit SizeEach