Stainless-Steel Spoons
Stainless-Steel Spoons
Actual product may vary
Bel-Art
Stainless-Steel Spoons
Double-ended spoons offer two bowl sizes for convenient sample handling
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Length (Metric)
FIS14-429E2 mL, 8 mL30 cm
FIS22-2615471.6 mL, 3 mL21 cm
Catalog number FIS14-429E
Price (SGD)
333.00
PK
Add to cart
Capacity (Metric):
2 mL, 8 mL
Length (Metric):
30 cm
Price (SGD)
333.00
PK
Add to cart
Double-ended spoons offer two bowl sizes for convenient sample handling
  • Polished 304 stainless-steel construction
  • Solid, rounded shaft for easy handling
Specifications
Product TypeSpoon
MaterialStainless Steel
AutoclavableAutoclavable
IncludesDouble end with 2mL small spoon at one end and 8mL large spoon at other
Capacity (Metric)2 mL, 8 mL
DisposableReusable
ColorSilver
Dimensions (L x W)11.81 x 1.5 in. (30 x 3.81 cm)
Length (English)11.81 in.
Length (English) Handle9.25 in.
Length (Metric)30 cm
Length (Metric) Handle23.49 cm
Width (English)1.5 in.
Width (Metric)3.81 cm
Unit SizePK