Bleach-Rite™ Test Strips
Actual product may vary
Current Technologies
Bleach-Rite™ Test Strips
Indicate if bleach solution meets the 0.525% minimum requirement for disinfecting hard surfaces and equipment
Have Questions?
Catalog number FIS14-412-60
Price (SGD)
-
  • Help determine if bleach disinfecting products are actually working. Bleach solutions can become non-active if the lid of a wipe canister is left open, if a bottle mixed in-house is weak / too old, or a bleach / water dual cartridge system is not spraying out a 1:10 solution
  • To use, dip one of the test strips into a sample of your bleach solution. The test strip will change color within 60 seconds; compare to the color chart on bottle. The color comparision warns if bleach solution is below the .525% (1:10 dilution) threshold for disinfection
  • Also help confirm if bleach solution is at the right concentration, so there is no need to throw it out. Don't waste time remixing if solution is strong enough
Specifications
For Use With (Application)Used to indicate if Bleach solution meets the 0.525% minimum requirement for disinfecting hard surfaces and equipment and for use on Glass, tile, laminated Plastic countertops
Test Time60 sec.
Unit SizeEach