Bio-Screen™ Biohazard Wipes, Liners, and Rolls
Actual product may vary
Bio-Screen™ Biohazard Wipes, Liners, and Rolls
Bio-Screen™ Biohazard Wipes, Liners, and Rolls
Current Technologies
Bio-Screen™ Biohazard Wipes, Liners, and Rolls
Low-linting, white, absorbent wipes bonded to an orange plastic backing
Have Questions?
Catalog NumberDimensions (L x W)Absorbancy
FIS14-412-5816 x 16 in. (406.4 x 406.4mm)High
Catalog number FIS14-412-58
Price (SGD)
413.00
Case
Dimensions (L x W):
16 x 16 in. (406.4 x 406.4mm)
Absorbancy:
High
Price (SGD)
413.00
Case
  • White absorbent side of this wipe / liner is low-linting and more absorbent than gauze or paper wipes
  • Orange plastic backing of wipe / liner blocks soak through of blood, body fluids, specimens and chemicals (xylene, ketones, formalin) to gloves and surfaces
  • Used for cleaning up small / large spills, to wipe pipets, probes, slide prep devices, and to pop caps from blood and specimen tubes
  • Limit cross-contamination from gloves as employees touch phones, keyboards, doors, etc.
  • Larger sizes can be used as mats and liners. Rolls of material can be cut to desired sizes
  • Three thicknesses available: Standard (absorbs 9 times its weight), Heavy (absorbs 12 times its weight), Super (absorbs 25 times its weight)
Wipes and Liners are non-sterile and not intended for use on patients.
Specifications
Product TypeBiohazard Wipe
AbsorbancyHigh
Length (English)16 in.
Length (Metric)406.4 mm
Width (English)16 in.
Width (Metric)406.4 mm
Dimensions (L x W)16 x 16 in. (406.4 x 406.4mm)
ColorWhite and Orange
For Use With (Application)Used for cleaning up small or large spills, to wipe pipets, probes, slide prep devices, and to pop caps from blood and specimen tubes. Wipes and Liners are non-sterile and not intended for use on patients.
QuantityEach
Unit SizeCase