HYPE-WIPE™ and Mini HYPE-WIPE™ Bleach Towelettes
Actual product may vary
HYPE-WIPE™ and Mini HYPE-WIPE™ Bleach Towelettes
HYPE-WIPE™ and Mini HYPE-WIPE™ Bleach Towelettes
HYPE-WIPE™ and Mini HYPE-WIPE™ Bleach Towelettes
HYPE-WIPE™ and Mini HYPE-WIPE™ Bleach Towelettes
HYPE-WIPE™ and Mini HYPE-WIPE™ Bleach Towelettes
Current Technologies
HYPE-WIPE™ and Mini HYPE-WIPE™ Bleach Towelettes
EPA approved to kill Clostridium difficile spores in four minutes
Have Questions?
Catalog NumberLength (English)Size
FIS14-412-5612 in.Standard
Catalog number FIS14-412-56
Price (SGD)
333.00
Case
Length (English):
12 in.
Size:
Standard
Price (SGD)
333.00
Case
Individually packaged, single-use towelettes saturated with 0.525% bleach (a 1:10 dilution). These bleach wipes do not dry out like other wipes will when their canister lid is left open.
 Registered as a hospital-grade cleaner/disinfectant with the U.S. EPA
  • Stabilized for 12-month shelf life with expiration date stamped on each foil pouch and outer box
  • Full size (6 in. x 12 in. towel folded inside a pocket-sized pouch) can wipe down 40 sq. ft. benchtop
  • Mini (3 in.x3 in.towel) is for cleaning glucometers, coagulation devices or small spills
  • For use on hard, non-porous surfaces including plastics, Plexiglas™, glass, glazed ceramic tile, linoleum, baked enamel, stainless steel (follow equipment manufacturers' directions)
  • Disinfection sites include POCT equipment, lab equipment, counters, centrifuges, cryostats, phlebotomy and specimen collection areas, wastewater treatment, pharma and research labs
Kill Time - One Minute
BacteriaViruses
Enterobacter cloacae NDM-1 Hepatitis B (HBV) 
Escherichia coli (ESBL)Hepatitis C (HCV)
Escherichia coli NDM-1Influenza A H1N1 (swine flu)
Klebsiella pneumoniae NDM-1 Rotavirus 
Carbapenem-Resistant Klebsiella pneumoniae (CRKP)Norovirus
Multi-drug Resistant (MDR) Enterococcus faeciumRespiratory Syncytial (RSV)
Multi-drug Resistant (MDR) Staphylococcus aureus 
Tuberculosis – TB (Mycobacterium bovis – BCG) 
Vancomycin-Resistant Staphylococcus aureus (VRSA) 
Pseudomonas aeruginosa 
Salmonella enterica 
Strepococcus pyogenes 
Kill Time - Two Minutes
Methicillin Resistant Staphylococcus aureus (MRSA)
Staphylococcus aureus
Vancomycin Resistant Enterococcus faecium (VRE)
Kill Time - Four Minutes
Clostridium difficile Spores (C. diff spores)
All kill claims, including Clostridium difficile Spore claim are registered with the Federal EPA and in the state of California and other states.
Specifications
Certifications/ComplianceEPA
Composition0.525% Bleach
Dilution0.048611111111
For Use With (Application)Disinfection of Blood Glucose monitors, counters, outer sides of equipment, IV poles, bed railings, probes, Blood pressure cuffs, slide prep areas, centrifuges, mammography equipment, carts, wheelchairs, gurneys, EMS backboards and phlebotomy trays
Length (English)12 in.
Length (Metric)30.5 cm
Shelf Life1 Year
SizeStandard
Unit SizeCase