Bleach-Rite™ Disinfecting Spray with Bleach
Actual product may vary
Bleach-Rite™ Disinfecting Spray with Bleach
Bleach-Rite™ Disinfecting Spray with Bleach
Bleach-Rite™ Disinfecting Spray with Bleach
Bleach-Rite™ Disinfecting Spray with Bleach
Current Technologies
Bleach-Rite™ Disinfecting Spray with Bleach
EPA certified to kill Clostridium difficile spores in four minutes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
FIS14-412-533.8 L
FIS14-412-65473 mL
Catalog number FIS14-412-53
Price (SGD)
373.00
Case
Volume (Metric):
3.8 L
Price (SGD)
373.00
Case
  • Bottles contain pre-mixed, ready-to-use bleach disinfectant formulated at 0.525% (a 1:10 dilution)
  • Stabilized for 18-month shelf life with expiration date on each bottle and outer box for ease of management/inspection
  • Disinfection sites include: POCT devices, lab equipment including chemistry/gas analyzers, counters, centrifuges, cryostats, phlebotomy and specimen collection areas, wastewater treatment, pharma and research labs and production areas
  • For use on hard, non-porous surfaces including plastics, Plexiglas™, glass, glazed ceramic tile, linoleum, baked enamel, stainless steel (follow equipment manufacturers' directions)
  • Available in 16 or 32 ounce bottles with sprayers; half gallon or gallon bottles
Kill Time - 30 Seconds
Bacteria
Vancomycin-resistant Enterococcus faecium (VRE)
Streptococcus pyogenes
Kill Time - One Minute
BacteriaVirusFungi
Acinetobacter baumanniiCoronavirus Aspergillus brasiliensis
Bordetella pertussisHepatitis A (HAV)Trichophyton mentagrophytes (Athlete's Foot Fungus)
Carbapenem-resistant Klebsiellapneumoniae (CRKP) Hepatitis B (HBV) 
Community acquired methicillin resistant staphylococcus aureus (CA-MRSA) Hepatitis C (HCV 
Enterobacter aerogenes Herpes Simplex (HSV-2) 
Escherichia coli (ESBL) HIV-1 (AIDS) 
Pseudomonas aeruginosa Influenza A H1N1 (swine flu) 
Salmonella enterica Influenza A 
Methicillin Resistant Staphylococcus aureus (MRSA) Norovirus 
Staphylococcus aureus  Poliovirus 
Vancomycin-resistant Staphylococcus aureus (VRSA) Respiratory Syncytial (RSV) 
  Rhinovirus 
 Rotavirus 
Kill Time - Two Minutes
TuberculocidalViruses
Mycobacterium bovis (BCG) TBCanine Parvovirus
 Feline Panleukopenia
Kill Time - Four Minutes
 Sporicidal
Clostridium difficile Spores (C. diff spores)
Specifications
Certifications/ComplianceEPA, NCCLS, OSHA, CDC
Composition0.525% Bleach
Container TypeBottle
Dilution0.048611111111
For Use With (Application)Disinfection of Blood Glucose monitors, counters, outer sides of equipment, IV poles, bed railings, probes, Blood pressure cuffs, slide prep areas, centrifuges, mammography equipment and many more
Shelf Life18 Months
Volume (English)128 oz. (1 gal.)
Volume (Metric)3.8 L
Unit SizeCase