Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Shakers
▸
Shaker Accessories
▸
Wrist Action™ Laboratory Shaker Accessory, Finger SUREGRIP™ Clamp
Actual product may vary
Burrell Scientific
Wrist Action™ Laboratory Shaker Accessory, Finger SUREGRIP™ Clamp
For use with Wrist Action Laboratory Shaker
Have Questions?
Catalog Number
For Use With (Equipment)
FIS14-400-140
Wrist Action Laboratory Shakers
Catalog number
FIS14-400-140
Price (SGD)
449.00
Each
Add to cart
For Use With (Equipment):
Wrist Action Laboratory Shakers
Price (SGD)
449.00
Each
Add to cart
Maximum adjustability and security
Three-prong vinyl-sleeved red grippers
Fully adjustable shaft to accommodate various vessels
Handle almost any shape flask, bottle or container up to 250mL
Material: Aluminum
Color: Silver and red
Specifications
Product Type
Clamp
For Use With (Equipment)
Wrist Action Laboratory Shakers
No. of Positions
2
Warranty and Service Offering
1 Year