Centrifuge Tube Mini-Cooler
Actual product may vary
BrandTech
Centrifuge Tube Mini-Cooler
Designed to protect a wide range of solutions (enzymes, DNA, RNA, cell suspensions) by helping to maintain freezer temperatures on the lab bench.
Have Questions?
Catalog NumberColorHolds Temp., Hours
FIS14-380-801Yellow-20°C for 60 min.
Catalog number FIS14-380-801
Price (SGD)
756.00
Each
Color:
Yellow
Holds Temp., Hours:
-20°C for 60 min.
Price (SGD)
756.00
Each
  • Manufactured from durable polycarbonate
  • Filled with non-toxic gel
  • Hold twelve 0.5 to 2.0 mL tubes
  • Provided individually
Specifications
For Use With (Equipment)Centrifuge tube
Holds Temp., Hours-20°C for 60 min.
MaterialPolycarbonate
Product TypeMini Cooler
ColorYellow
Capacity (Metric)0.5 to 2 mL
Holds0.5 to 2 mL Tubes
AutoclavableNot Autoclavable
Temperature ControlPassive
Temperature Range-20°C
Unit SizeEach