PFA Round Bottom Flasks
Actual product may vary
BrandTech
PFA Round Bottom Flasks
Corrosion- and leach-resistant flasks excellent for rotary evaporation and distillation of high-purity chemicals
Have Questions?
Catalog NumberCapacity (Metric)
FIS14-379-201500 mL
Catalog number FIS14-379-201
Price (SGD)
1,019.00
Each
Capacity (Metric):
500 mL
Price (SGD)
1,019.00
Each
  • Manufactured from PFA (a perfluoroalkoxy polymer) for excellent heat and chemical resistance
  • Smooth surfaces simplifies cleaning and reduces risk of sample carryover in trace analysis applications
  • Maximum operating temperature under vacuum at 50°C (122°F)
  • Translucent surfaces for easy solvent monitoring
  • Ideal for rotary evaporation and distillation of high-purity chemicals
  • Specifications
    Capacity (Metric)500 mL
    MaterialPFA
    For Use With (Application)For rotary evaporation and distillation of high-purity chemicals
    IncludesGround socket
    Product TypeDistilling Flask
    AutoclavableAutoclavable
    DisposableReusable
    GraduatedNo
    Mouth StyleJoint
    SterilityNon-sterile
    Bottom ShapeRound
    Unit SizeEach