Graduated PMP Cylinders
Actual product may vary
BrandTech
Graduated PMP Cylinders
VITLAB™ polymethylpentene graduated cylinders, certified Class A to DIN 12681 and ISO 6706
Have Questions?
Catalog NumberCapacity (Metric)
FIS14-379-142100 mL
Catalog number FIS14-379-142
Price (SGD)
106.00
Pack
Capacity (Metric):
100 mL
Price (SGD)
106.00
Pack
  • Molded graduations with ring marks at major increments
  • Imprinted lot and year
  • Supplied certificate includes mean test value and lot
Specifications
Capacity (Metric)100 mL
MaterialPMP
Tolerance±0.5 mL
Certifications/ComplianceDIN 12681, ISO 6706
ClassClass A
CalibrationTo Contain
Diameter (English)1.29 in.
Diameter (Metric)33 mm
Graduations1 mL
AutoclavableAutoclavable
Subdivision1mL
ShapeRound
Base TypeHexagonal
Height (English)9.84 in.
Height (Metric)250 mm
Product TypeGraduated Cylinder
Unit SizePack