Polypropylene Griffin Beakers
Actual product may vary
BrandTech
Polypropylene Griffin Beakers
Polypropylene Griffin Beakers are embossed with easily readable raised blue scale for accurate measurement.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS14-379-029500 mL
FIS14-379-020100 mL
FIS14-379-024250 mL
FIS14-379-0191000 mL
Catalog number FIS14-379-029
Price (SGD)
125.00
Pack
Capacity (Metric):
500 mL
Price (SGD)
125.00
Pack
Manufactured with highly transparent polypropylene for optimum visibility.
  • To preserve markings, do not clean at temperatures exceeding 60°C
  • Conditionally autoclavable at 121°C, for design with raised graduations
  • Suitable for foodstuff
  • Manufactured in accordance with ISO 7056
Specifications
ColorTranslucent
Diameter (English) Outer3.34 in.
Diameter (Metric) Outer8.5 cm
Capacity (Metric)500 mL
MaterialPolypropylene
For Use With (Application)For General Purpose Use
Height (English)4.62 in.
Height (Metric)11.75 cm
Increments100 mL
Temperature (English) Max. Operating257°F
Temperature (Metric) Max. Operating125°C
TypeBeaker
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
Certifications/ComplianceISO 7056
ScaleMolded and Printed Graduated Scale
StyleGriffin
Unit SizePack