Pipet Keeper™ Biohazard Container
Actual product may vary
Whitney Medical Solutions
Pipet Keeper™ Biohazard Container
Disposable containers save space, time, and money
Have Questions?
Catalog NumberDimensions (L x W x H)
FIS14-375-26815.5 x 3 x 3 in.
Catalog number FIS14-375-268
Price (SGD)
568.00
Case
Dimensions (L x W x H):
15.5 x 3 x 3 in.
Price (SGD)
568.00
Case
  • Space-saving, economical, disposable point-of-use contaminated waste containers for disposal of pipets and other objects up to 15 in. (38.1cm) in length
  • Made of sturdy, plastic-lined paperboard with webbed corners to contain liquid
  • Interlocking top with reclosable flap
  • Folds out to 3W × 3H × 15.5 in.L (7.6 × 7.6 × 38.1cm) and accommodates up to 2100cc of material
  • Autoclavable
  • Made in USA
Specifications
DesignPrinted Biohazard Symbols
Capacity (English)2250 cc
Dimensions (L x W x H)15.5 x 3 x 3 in.
DisposableSingle-use
Height (Metric)7.6 cm
Length (Metric)39.4 cm
Width (Metric)7.6 cm
AutoclavableAutoclavable
IncludesSelf-locking top
Liner MaterialPlastic
For Use With (Application)Storing contaminated pipets up to 15 in. L
Width (English)3 in.
Length (English)15.5 in.
Height (English)3 in.
MaterialPaperboard
Product TypePipet Keeper
Unit SizeCase