Check Order Status
100-Slide Capacity Microscope Slide Storage Boxes
Actual product may vary
100-Slide Capacity Microscope Slide Storage Boxes
Designed for safe, long-term slide storage
Have Questions?
Change view
Catalog Number
Color
FIS14-372-87
Yellow
FIS14-372-85
Red
FIS14-372-86
Blue
FIS14-372-88
White
4 Options
Catalog number
FIS14-372-87
Price (SGD)
-
Color:
Yellow
Constructed from polystyrene
100-Slide capacity
Corrosion-resistant hinges and clasp
Cork-lined bottoms
Individually numbered slide positions
Available in four colors
Specifications
Slide Capacity
100
Color
Yellow
Device Holds
100
For Use With (Equipment)
Microscope Slide
Includes
Cork lined bottom
Material
Polystyrene