Handi-Hold™ Spatula
Handi-Hold™ Spatula
Actual product may vary
Handi-Hold™ Spatula
Fisherbrand Handi-Hold™ Spatulas are specially designed with a soft-grip handle for maximized comfort. Our Handi-Hold™ Spatulas are sturdy, economical tools for handling, transferring, blending, or collecting chemical and biological samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric) Blade
FIS14-366E20.32 cm
FIS14-366B10.16 cm
FIS14-366D15.24 cm
Catalog number FIS14-366E
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric) Blade:
20.32 cm
  • Fisherbrand™ Handi-Hold™ Spatula are specially designed with a soft-grip handle for maximized comfort.
  • Sturdy, economical tools for handling, transferring, blending, or collecting chemical and biological samples
  • Multiple sizes allow for varied laboratory applications
  • Resists deterioration by boiling water, acids, and most solvents
  • Maximized comfort with pvc soft-grip handles
  • Multipurpose - Useful in educational, research or healthcare setting
  • Made from ASTM standard (F899-94) series 400 stainless
  • Non-sticking blades
  • Electronically passivated for corrosion resistance
  • Can be sterilized by Eto/gamma radiation
  • Rockwell hardness (HRC): 40-45
  • Autoclavable up to 135°C
  • Latex-free
Specifications
Product TypeSpatula, Handi-Hold™
Length (English) Blade8 in.
Length (Metric) Blade20.32 cm
Blade MaterialStainless Steel
Handle MaterialPVC
Unit SizeEA